ADVH14DBC7C9 AChemBlock

1-Ethynyl-4-(phenylmethoxy)benzene

  • MDL number MFCD01814885
  • Cas Number : 84284-70-8
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-ethynyl-4-phenylmethoxy-benzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35860
Availability details for
Quantity
  • Availability
  • Add to Cart