ADVH14DBC7CD AChemBlock

5-Ethynyl-1,2,3-trifluorobenzene

  • MDL number MFCD08458177
  • Cas Number : 158816-55-8
  • Molecular Weight : 156.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-ethynyl-1-2-3-trifluorobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40896
Availability details for
Quantity
  • Availability
  • Add to Cart