ADVH14DBC7E4 AChemBlock

4-(3-Aminophenyl)-2-methyl-3-butyn-2-ol

  • MDL number MFCD06200785
  • Cas Number : 69088-96-6
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-3-aminophenyl-2-methyl-3-butyn-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36581
Availability details for
Quantity
  • Availability
  • Add to Cart