ADVH14DBC855 AChemBlock

(3R)-3-Hydroxy-2-piperidinone

  • MDL number MFCD11112158
  • Cas Number : 220928-01-8
  • Molecular Weight : 115.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3r-3-hydroxy-2-piperidinone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39619
Availability details for
Quantity
  • Availability
  • Add to Cart