ADVH14DBC87A AChemBlock

4-Bromo-3-chlorobenzeneacetic acid

  • MDL number MFCD18390583
  • Cas Number : 1261643-24-6
  • Molecular Weight : 249.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-3-chlorobenzeneacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41923
Availability details for
Quantity
  • Availability
  • Add to Cart