ADVH14DBC889 AChemBlock

3-Bromo-4-phenoxybenzaldehyde

  • MDL number MFCD27937159
  • Cas Number : 1000414-11-8
  • Molecular Weight : 277.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-phenoxybenzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43142
Availability details for
Quantity
  • Availability
  • Add to Cart