ADVH14DBC8AF AChemBlock

4-(4-Bromophenoxy)-1-butanol

  • MDL number MFCD12186897
  • Cas Number : 60222-62-0
  • Molecular Weight : 245.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-bromophenoxy-1-butanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37159
Availability details for
Quantity
  • Availability
  • Add to Cart