ADVH14DBC8C4 AChemBlock

N-Boc 6-(methylamino)hexan-1-ol

  • MDL number MFCD30533780
  • Cas Number : 1338075-62-9
  • Molecular Weight : 231.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-6-methylamino-hexan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41617
Availability details for
Quantity
  • Availability
  • Add to Cart