ADVH14DBC8DB AChemBlock

tert-Butyl 4-formylbenzoate

  • MDL number MFCD01111994
  • Cas Number : 65874-27-3
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-formylbenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36777
Availability details for
Quantity
  • Availability
  • Add to Cart