ADVH14DBC8FF AChemBlock

3'-Fluoro-biphenyl-3-carboxaldehyde

  • MDL number MFCD03424620
  • Cas Number : 400750-09-6
  • Molecular Weight : 200.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-biphenyl-3-carboxaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38274
Availability details for
Quantity
  • Availability
  • Add to Cart