ADVH14DBC906 AChemBlock

2'-Chloro-biphenyl-2-carboxaldehyde

  • MDL number MFCD04039056
  • Cas Number : 223575-76-6
  • Molecular Weight : 216.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-biphenyl-2-carboxaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39581
Availability details for
Quantity
  • Availability
  • Add to Cart