ADVH14DBC915 AChemBlock

4-(5-Formylfuran-2-yl)benzoic acid

  • MDL number MFCD00455087
  • Cas Number : 39245-15-3
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-5-formylfuran-2-yl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38348
Availability details for
Quantity
  • Availability
  • Add to Cart