ADVH14DBC921 AChemBlock

2,2-Bis(phenylmethyl)propanedioic acid

  • MDL number MFCD03684094
  • Cas Number : 4372-32-1
  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-bis-phenylmethyl-propanedioic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38086
Availability details for
Quantity
  • Availability
  • Add to Cart