ADVH14DBC937 AChemBlock

1-Boc-3-(methylsulfonyloxy)pyrrolidine

  • MDL number MFCD04116219
  • Cas Number : 141699-57-2
  • Molecular Weight : 265.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-methylsulfonyloxy-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41337
Availability details for
Quantity
  • Availability
  • Add to Cart