ADVH14DBC948 AChemBlock

1-Boc-3-(tosyloxy-methyl)pyrrolidine

  • MDL number MFCD08704608
  • Cas Number : 114214-70-9
  • Molecular Weight : 355.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-tosyloxy-methyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42569
Availability details for
Quantity
  • Availability
  • Add to Cart