ADVH14DBC958 AChemBlock

3-(2-Fluorophenoxy)pyrrolidine hydrochloride

  • MDL number MFCD18459178
  • Cas Number : 1211507-42-4
  • Molecular Weight : 217.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-fluorophenoxy-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42281
Availability details for
Quantity
  • Availability
  • Add to Cart