ADVH14DBC96E AChemBlock

2-[2-(Trifluoromethyl)phenyl]pyrrolidine hydrochloride

  • MDL number MFCD03840006
  • Cas Number : 524674-04-2
  • Molecular Weight : 251.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-trifluoromethyl-phenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37638
Availability details for
Quantity
  • Availability
  • Add to Cart