ADVH14DBC976 AChemBlock

2-(Benzodioxol-5-yl)pyrrolidine hydrochloride

  • MDL number MFCD31560332
  • Cas Number : 95849-38-0
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-benzodioxol-5-yl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34808
Availability details for
Quantity
  • Availability
  • Add to Cart