ADVH14DBC98A AChemBlock

3-[4-(Trifluoromethoxy)phenoxy]pyrrolidine hydrochloride

  • MDL number MFCD31560309
  • Cas Number : 946715-68-0
  • Molecular Weight : 283.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-trifluoromethoxy-phenoxy-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34900
Availability details for
Quantity
  • Availability
  • Add to Cart