ADVH14DBC9B2 AChemBlock

4-Piperidinepropanol

  • MDL number MFCD02178380
  • Cas Number : 7037-49-2
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-piperidinepropanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36532
Availability details for
Quantity
  • Availability
  • Add to Cart