ADVH14DBC9CB AChemBlock

N-Boc-3-bromo-4-piperidone

  • MDL number MFCD06738530
  • Cas Number : 188869-05-8
  • Molecular Weight : 278.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-3-bromo-4-piperidone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40224
Availability details for
Quantity
  • Availability
  • Add to Cart