ADVH14DBC9D4 AChemBlock

1-Cbz-4-(mesyloxy)piperidine

  • MDL number MFCD09953312
  • Cas Number : 199103-19-0
  • Molecular Weight : 313.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-cbz-4-mesyloxy-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40053
Availability details for
Quantity
  • Availability
  • Add to Cart