ADVH14DBC9DD AChemBlock

1-Boc-4-piperidineacetic acid methyl ester

  • MDL number MFCD01632529
  • Cas Number : 175213-46-4
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-piperidineacetic-acid-methyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40502
Availability details for
Quantity
  • Availability
  • Add to Cart