ADVH14DBC9EA AChemBlock

Methyl 2-amino-2-phenylpropanoate hydrochloride

  • MDL number MFCD16817459
  • Cas Number : 191796-90-4
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-amino-2-phenylpropanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40173
Availability details for
Quantity
  • Availability
  • Add to Cart