ADVH14DBCA5E AChemBlock

a-(Trifluoromethyl)-1H-indole-3-ethanamine

  • MDL number MFCD18909182
  • Cas Number : 137496-38-9
  • Molecular Weight : 228.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-trifluoromethyl-1h-indole-3-ethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41474
Availability details for
Quantity
  • Availability
  • Add to Cart