ADVH14DBCA61 AChemBlock

3-Chlorobenzamide

  • MDL number MFCD00014799
  • Cas Number : 618-48-4
  • Molecular Weight : 155.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chlorobenzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37042
Availability details for
Quantity
  • Availability
  • Add to Cart