ADVH14DBCA62 AChemBlock

2,2-Difluoro-N-methylethanamine hydrochloride

  • MDL number MFCD11043069
  • Cas Number : 139364-36-6
  • Molecular Weight : 131.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluoro-n-methylethanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41399
Availability details for
Quantity
  • Availability
  • Add to Cart