ADVH14DBCA92 AChemBlock

2-Chloro-4-[(methylamino)methyl]pyridine

  • MDL number MFCD06654462
  • Cas Number : 748187-76-0
  • Molecular Weight : 156.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-4-methylamino-methyl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36290
Availability details for
Quantity
  • Availability
  • Add to Cart