ADVH14DBCA98 AChemBlock

4-Methoxy-3-oxo-butanoic acid benzyl ester

  • MDL number MFCD11849518
  • Cas Number : 242486-38-0
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methoxy-3-oxo-butanoic-acid-benzyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39432
Availability details for
Quantity
  • Availability
  • Add to Cart