ADVH14DBCAFF AChemBlock

(R)-Methyl 3-amino-4-(2,5-difluorophenyl)butanoate hydrochloride

  • MDL number MFCD31699831
  • Cas Number : 848667-53-8
  • Molecular Weight : 265.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-methyl-3-amino-4-2-5-difluorophenyl-butanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35824
Availability details for
Quantity
  • Availability
  • Add to Cart