ADVH14DBCB20 AChemBlock

2-Amino-N-isopropylacetamide

  • MDL number MFCD07310996
  • Cas Number : 67863-05-2
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-n-isopropylacetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36647
Availability details for
Quantity
  • Availability
  • Add to Cart