ADVH14DBCB44 AChemBlock

(3S)-1-Methyl-3-pyrrolidinamine dihydrochloride

  • MDL number MFCD17010247
  • Cas Number : 874348-06-8
  • Molecular Weight : 173.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3s-1-methyl-3-pyrrolidinamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35579
Availability details for
Quantity
  • Availability
  • Add to Cart