ADVH14DBCB4F AChemBlock

(2S)-2-(Fmoc-amino)tetradecanoic acid

  • MDL number MFCD08275903
  • Cas Number : 478706-50-2
  • Molecular Weight : 465.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2s-2-fmoc-amino-tetradecanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37920
Availability details for
Quantity
  • Availability
  • Add to Cart