ADVH14DBCB60 AChemBlock

N-Fmoc-N-hexyl-glycine

  • MDL number MFCD30187633
  • Cas Number : 1374785-50-8
  • Molecular Weight : 381.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-fmoc-n-hexyl-glycine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41475
Availability details for
Quantity
  • Availability
  • Add to Cart