ADVH14DBCB61 AChemBlock

N-Fmoc-N-(2-phenylethyl)-glycine

  • MDL number MFCD30187632
  • Cas Number : 540483-58-7
  • Molecular Weight : 401.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-fmoc-n-2-phenylethyl-glycine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37552
Availability details for
Quantity
  • Availability
  • Add to Cart