ADVH14DBCB72 AChemBlock

1-Acetyl-4-piperidinecarboxaldehyde

  • MDL number MFCD11582881
  • Cas Number : 155826-26-9
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-acetyl-4-piperidinecarboxaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40953
Availability details for
Quantity
  • Availability
  • Add to Cart