ADVH14DBCB73 AChemBlock

(4-Fluorophenyl)-3-piperidinyl-methanone hydrochloride

  • MDL number MFCD11846283
  • Cas Number : 118412-66-1
  • Molecular Weight : 243.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluorophenyl-3-piperidinyl-methanone-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42428
Availability details for
Quantity
  • Availability
  • Add to Cart