ADVH14DBCBBF AChemBlock

1-Fmoc-2-methyl-DL-proline

  • MDL number MFCD20304802
  • Cas Number : 433220-67-8
  • Molecular Weight : 351.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-fmoc-2-methyl-dl-proline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38100
Availability details for
Quantity
  • Availability
  • Add to Cart