ADVH14DBCBC9 AChemBlock

(S)-3-Amino-5-methylhexanoic acid

  • MDL number MFCD01076258
  • Cas Number : 22818-43-5
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-amino-5-methylhexanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39532
Availability details for
Quantity
  • Availability
  • Add to Cart