ADVH14DBCBEB AChemBlock

3-(4-Methoxyphenyl)-2-propynoic acid ethyl ester

  • MDL number MFCD07772987
  • Cas Number : 51718-85-5
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-methoxyphenyl-2-propynoic-acid-ethyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37696
Availability details for
Quantity
  • Availability
  • Add to Cart