ADVH14DBCBF4 AChemBlock

3-[[2-(2-Methoxyethoxy)ethoxy]methyl]-3-methyloxetane

  • MDL number MFCD30531305
  • Cas Number : 531521-23-0
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-2-methoxyethoxy-ethoxy-methyl-3-methyloxetane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37601
Availability details for
Quantity
  • Availability
  • Add to Cart