ADVH14DBCC06 AChemBlock

2-Methyl-a-oxo-benzeneacetic acid

  • MDL number MFCD07698691
  • Cas Number : 150781-72-9
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-a-oxo-benzeneacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41054
Availability details for
Quantity
  • Availability
  • Add to Cart