ADVH14DBCC0E AChemBlock

10-Bromo-decanoic acid ethyl ester

  • MDL number MFCD01112172
  • Cas Number : 55099-31-5
  • Molecular Weight : 279.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/10-bromo-decanoic-acid-ethyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37482
Availability details for
Quantity
  • Availability
  • Add to Cart