ADVH14DBCC0F AChemBlock

9-Bromo-nonanoic acid methyl ester

  • MDL number MFCD21337473
  • Cas Number : 67878-15-3
  • Molecular Weight : 251.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/9-bromo-nonanoic-acid-methyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36645
Availability details for
Quantity
  • Availability
  • Add to Cart