ADVH14DBCC37 AChemBlock

2'-Chloro-biphenyl-4-carboxaldehyde

  • MDL number MFCD01631914
  • Cas Number : 39802-78-3
  • Molecular Weight : 216.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-biphenyl-4-carboxaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38318
Availability details for
Quantity
  • Availability
  • Add to Cart