ADVH14DBCC5E AChemBlock

Methyl 5-(bromomethyl)-2-chlorobenzoate

  • MDL number MFCD20483458
  • Cas Number : 16220-99-8
  • Molecular Weight : 263.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromomethyl-2-chlorobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40803
Availability details for
Quantity
  • Availability
  • Add to Cart