ADVH14DBCC7F AChemBlock

6-Benzyloxy-1-hexanol

  • MDL number MFCD03265790
  • Cas Number : 71126-73-3
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-benzyloxy-1-hexanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36477
Availability details for
Quantity
  • Availability
  • Add to Cart