ADVH14DBCCCF AChemBlock

(S)-3-Cbz-4-methyl-1,2,3-oxathiazolidine 2,2-dioxide

  • MDL number MFCD31568494
  • Cas Number : 1007121-72-3
  • Molecular Weight : 271.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-cbz-4-methyl-1-2-3-oxathiazolidine-2-2-dioxide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43105
Availability details for
Quantity
  • Availability
  • Add to Cart