ADVH14DBCCF3 AChemBlock

Benzyl (R)-Lactate

  • MDL number MFCD00672497
  • Cas Number : 74094-05-6
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzyl-r-lactate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36350
Availability details for
Quantity
  • Availability
  • Add to Cart