ADVH14DBCCFD AChemBlock

4-Fluoro-3-nitro-benzamide

  • MDL number MFCD11110229
  • Cas Number : 349-02-0
  • Molecular Weight : 184.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-3-nitro-benzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38641
Availability details for
Quantity
  • Availability
  • Add to Cart